siRNA Details
| Gene Symbol: Lzic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013241.1 | NP_001013259.1 | 3 | 465 | ||
| XM_006239434.2 | XP_006239496.1 | 3 | 607 | |||
| XM_006239435.1 | XP_006239497.1 | 3 | 209 | |||
| GenBank mRNA | BC087098.1 | - | 3 | 465 | 7191966 | |

Lzic
Genomic Map