siRNA Details
| Gene Symbol: Pik3cd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108978.1 | NP_001102448.1 | 13 | 1873 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Pik3cd
Pik3cd| Gene Symbol: Pik3cd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108978.1 | NP_001102448.1 | 13 | 1873 | ||
 Back To Top
 Back To Top