siRNA Details
| Gene Symbol: Pik3cd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108978.1 | NP_001102448.1 | 20 | 2836 | ||
| GenBank mRNA | BC099153.1 | - | 3 | 1563 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pik3cd| Gene Symbol: Pik3cd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108978.1 | NP_001102448.1 | 20 | 2836 | ||
| GenBank mRNA | BC099153.1 | - | 3 | 1563 | ||