siRNA Details
| Gene Symbol: Wrap73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014262.1 | NP_001014284.1 | 3, 4 | 362 | ||
| GenBank mRNA | BC082062.1 | - | 3, 4 | 362 | 7132552 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wrap73| Gene Symbol: Wrap73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014262.1 | NP_001014284.1 | 3, 4 | 362 | ||
| GenBank mRNA | BC082062.1 | - | 3, 4 | 362 | 7132552 | |