siRNA Details
| Gene Symbol: Slc30a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013243.1 | NP_001013261.1 | 1, 2 | 177 | ||
| GenBank mRNA | AY538655.1 | - | 1, 2 | 177 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc30a3| Gene Symbol: Slc30a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013243.1 | NP_001013261.1 | 1, 2 | 177 | ||
| GenBank mRNA | AY538655.1 | - | 1, 2 | 177 | ||