siRNA Details
| Gene Symbol: Dtd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108981.1 | NP_001102451.1 | 3 | 217 | ||
| GenBank mRNA | BC159434.1 | - | 212 | 9100851 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dtd2| Gene Symbol: Dtd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108981.1 | NP_001102451.1 | 3 | 217 | ||
| GenBank mRNA | BC159434.1 | - | 212 | 9100851 | ||