siRNA Details
| Gene Symbol: LOC366772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY158661.1 | - | 338 | |||
| Gene Symbol: Igh-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC092586.1 | - | 305 | 7366480 | ||

LOC366772
Genomic Map