siRNA Details
| Gene Symbol: Olr921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001385.1 | NP_001001385.1 | 1 | 896 | ||
| Gene Symbol: LOC103689923 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765069.1 | XP_008763291.1 | 1 | 896 | ||
| Gene Symbol: Olr903 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001361.1 | NP_001001361.1 | 1 | 896 | ||

Olr921
Genomic Map