siRNA Details
| Gene Symbol: Olr932 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001386.1 | NP_001001386.1 | 1 | 881 | ||
| Gene Symbol: Olr1052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001363.1 | NP_001001363.1 | 1 | 881 | ||

Olr932
Genomic Map