siRNA Details
| Gene Symbol: Olr1070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000591.1 | NP_001000591.1 | 1 | 124 | ||
| Gene Symbol: Olr1071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000063.1 | NP_001000063.1 | 1 | 124 | ||

Olr1070
Genomic Map