siRNA Details
| Gene Symbol: Olr1093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001388.1 | NP_001001388.1 | 1 | 758 | ||
| Gene Symbol: Olr1090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001379.1 | NP_001001379.1 | 758 | |||
| Gene Symbol: Olr1092 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001380.1 | NP_001001380.1 | 1 | 758 | ||
More Information
Set Membership: |
|

Olr1093
Genomic Map