siRNA Details
| Gene Symbol: Kank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108989.1 | NP_001102459.1 | 6, 7 | 1765 | ||
| GenBank mRNA | BC166847.1 | - | 6, 7 | 1748 | 9030846 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kank3| Gene Symbol: Kank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108989.1 | NP_001102459.1 | 6, 7 | 1765 | ||
| GenBank mRNA | BC166847.1 | - | 6, 7 | 1748 | 9030846 | |