siRNA Details
| Gene Symbol: Kank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108989.1 | NP_001102459.1 | 3 | 1293 | ||
| XM_008765202.1 | XP_008763424.1 | 3 | 1320 | |||
| GenBank mRNA | BC166847.1 | - | 3 | 1276 | 9030846 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kank3| Gene Symbol: Kank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108989.1 | NP_001102459.1 | 3 | 1293 | ||
| XM_008765202.1 | XP_008763424.1 | 3 | 1320 | |||
| GenBank mRNA | BC166847.1 | - | 3 | 1276 | 9030846 | |