siRNA Details
| Gene Symbol: Cep83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014266.1 | NP_001014288.1 | 4 | 486 | ||
| XM_008765317.1 | XP_008763539.1 | 4 | 482 | |||
| XM_008765318.1 | XP_008763540.1 | 3 | 328 | |||
| GenBank mRNA | BC081969.1 | - | 4 | 486 | 7125504 | |

Cep83
Genomic Map