siRNA Details
| Gene Symbol: Zdhhc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039340.1 | NP_001034429.1 | 11, 12 | 1252 | ||
| GenBank mRNA | AY886533.1 | - | 11, 12 | 1252 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zdhhc17| Gene Symbol: Zdhhc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039340.1 | NP_001034429.1 | 11, 12 | 1252 | ||
| GenBank mRNA | AY886533.1 | - | 11, 12 | 1252 | ||