siRNA Details
| Gene Symbol: Caps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108990.1 | NP_001102460.1 | 4 | 330 | ||
| XM_006241337.2 | XP_006241399.1 | 14 | 1678 | |||
| XM_008765359.1 | XP_008763581.1 | 13 | 1210 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Caps2| Gene Symbol: Caps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108990.1 | NP_001102460.1 | 4 | 330 | ||
| XM_006241337.2 | XP_006241399.1 | 14 | 1678 | |||
| XM_008765359.1 | XP_008763581.1 | 13 | 1210 | |||