siRNA Details
| Gene Symbol: Caps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108990.1 | NP_001102460.1 | 7 | 668 | ||
| XM_006241337.2 | XP_006241399.1 | 17 | 2016 | |||
| XM_008765359.1 | XP_008763581.1 | 16 | 1548 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Caps2| Gene Symbol: Caps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108990.1 | NP_001102460.1 | 7 | 668 | ||
| XM_006241337.2 | XP_006241399.1 | 17 | 2016 | |||
| XM_008765359.1 | XP_008763581.1 | 16 | 1548 | |||