siRNA Details
| Gene Symbol: Trhde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108991.1 | NP_001102461.1 | 2, 3 | 1803 | ||
| GenBank mRNA | X80535.1 | - | 1, 2 | 862 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trhde| Gene Symbol: Trhde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108991.1 | NP_001102461.1 | 2, 3 | 1803 | ||
| GenBank mRNA | X80535.1 | - | 1, 2 | 862 | ||