siRNA Details
| Gene Symbol: Cpsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130571.1 | NP_001124043.1 | 5 | 608 | ||
| XM_006241853.2 | XP_006241915.1 | 5 | 603 | |||
| GenBank mRNA | BC168713.1 | - | 5 | 516 | 8367087 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpsf1| Gene Symbol: Cpsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130571.1 | NP_001124043.1 | 5 | 608 | ||
| XM_006241853.2 | XP_006241915.1 | 5 | 603 | |||
| GenBank mRNA | BC168713.1 | - | 5 | 516 | 8367087 | |