siRNA Details
| Gene Symbol: Tonsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130572.1 | NP_001124044.1 | 18 | 2858 | ||
| XM_008765587.1 | XP_008763809.1 | 18 | 2906 | |||
| XM_008765588.1 | XP_008763810.1 | 18 | 2906 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tonsl| Gene Symbol: Tonsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130572.1 | NP_001124044.1 | 18 | 2858 | ||
| XM_008765587.1 | XP_008763809.1 | 18 | 2906 | |||
| XM_008765588.1 | XP_008763810.1 | 18 | 2906 | |||