siRNA Details
| Gene Symbol: Zfp251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191911.1 | NP_001178840.1 | 4 | 1854 | ||
| XM_006241854.1 | XP_006241916.1 | 3 | 1738 | |||
| XM_008765589.1 | XP_008763811.1 | 2 | 1673 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp251| Gene Symbol: Zfp251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191911.1 | NP_001178840.1 | 4 | 1854 | ||
| XM_006241854.1 | XP_006241916.1 | 3 | 1738 | |||
| XM_008765589.1 | XP_008763811.1 | 2 | 1673 | |||