siRNA Details
| Gene Symbol: Tcf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130574.1 | NP_001124046.1 | 2 | 2432 | ||
| XM_006242110.2 | XP_006242172.1 | 1 | 3332 | |||
| XM_006242112.2 | XP_006242174.1 | 1 | 3332 | |||
| XM_008765772.1 | XP_008763994.1 | 1 | 3331 | |||

Tcf20
Genomic Map