siRNA Details
| Gene Symbol: Tbc1d22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242171.2 | XP_006242233.1 | 4, 5 | 2453 | ||
| XM_008765824.1 | XP_008764046.1 | 4, 5 | 2451 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d22a| Gene Symbol: Tbc1d22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242171.2 | XP_006242233.1 | 4, 5 | 2453 | ||
| XM_008765824.1 | XP_008764046.1 | 4, 5 | 2451 | |||