siRNA Details
| Gene Symbol: Pphln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108992.1 | NP_001102462.1 | 3 | 470 | ||
| XM_006242230.1 | XP_006242292.1 | 3 | 467 | |||
| XM_006242231.1 | XP_006242293.1 | 3 | 467 | |||
| XM_006242236.2 | XP_006242298.1 | 3 | 234 | |||
| GenBank mRNA | AY724510.1 | - | 2 | 203 | ||

Pphln1
Genomic Map