siRNA Details
| Gene Symbol: Tbca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013245.1 | NP_001013263.1 | 2 | 98 | ||
| XM_008760682.1 | XP_008758904.1 | 2 | 134 | |||
| GenBank mRNA | BC058155.1 | - | 2 | 98 | 6888541 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbca| Gene Symbol: Tbca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013245.1 | NP_001013263.1 | 2 | 98 | ||
| XM_008760682.1 | XP_008758904.1 | 2 | 134 | |||
| GenBank mRNA | BC058155.1 | - | 2 | 98 | 6888541 | |