siRNA Details
| Gene Symbol: Olr1230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000595.1 | NP_001000595.1 | 1 | 160 | ||
| XM_008762652.1 | XP_008760874.1 | 3 | 350 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1230| Gene Symbol: Olr1230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000595.1 | NP_001000595.1 | 1 | 160 | ||
| XM_008762652.1 | XP_008760874.1 | 3 | 350 | |||