siRNA Details
| Gene Symbol: Arhgef12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013246.1 | NP_001013264.1 | 20 | 1692 | ||
| GenBank mRNA | AY935257.1 | - | 20 | 1692 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef12| Gene Symbol: Arhgef12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013246.1 | NP_001013264.1 | 20 | 1692 | ||
| GenBank mRNA | AY935257.1 | - | 20 | 1692 | ||