siRNA Details
| Gene Symbol: Arhgap20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213629.1 | NP_998794.1 | 15 | 2064 | ||
| XM_006243047.2 | XP_006243109.1 | 15 | 2075 | |||
| GenBank mRNA | AY501475.1 | - | 15 | 2064 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap20| Gene Symbol: Arhgap20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213629.1 | NP_998794.1 | 15 | 2064 | ||
| XM_006243047.2 | XP_006243109.1 | 15 | 2075 | |||
| GenBank mRNA | AY501475.1 | - | 15 | 2064 | ||