siRNA Details
| Gene Symbol: LOC367117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047919.1 | NP_001041384.1 | 2 | 160 | ||
| GenBank mRNA | AY539938.1 | - | 2 | 160 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC367117| Gene Symbol: LOC367117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047919.1 | NP_001041384.1 | 2 | 160 | ||
| GenBank mRNA | AY539938.1 | - | 2 | 160 | ||