siRNA Details
| Gene Symbol: Hyal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207616.1 | NP_997499.1 | 2 | 121 | ||
| GenBank mRNA | AB100600.1 | - | 2 | 121 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hyal1| Gene Symbol: Hyal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207616.1 | NP_997499.1 | 2 | 121 | ||
| GenBank mRNA | AB100600.1 | - | 2 | 121 | ||