siRNA Details
| Gene Symbol: Traip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109004.2 | NP_001102474.1 | 7, 8 | 691 | ||
| XM_008766558.1 | XP_008764780.1 | 7, 8 | 1300 | |||
| GenBank mRNA | BC162006.1 | - | 7, 8 | 691 | 9089897 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Traip| Gene Symbol: Traip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109004.2 | NP_001102474.1 | 7, 8 | 691 | ||
| XM_008766558.1 | XP_008764780.1 | 7, 8 | 1300 | |||
| GenBank mRNA | BC162006.1 | - | 7, 8 | 691 | 9089897 | |