siRNA Details
| Gene Symbol: Traip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109004.2 | NP_001102474.1 | 13, 14 | 1315 | ||
| GenBank mRNA | BC162006.1 | - | 13, 14 | 1315 | 9089897 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Traip| Gene Symbol: Traip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109004.2 | NP_001102474.1 | 13, 14 | 1315 | ||
| GenBank mRNA | BC162006.1 | - | 13, 14 | 1315 | 9089897 | |