siRNA Details
| Gene Symbol: Dhx30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013249.1 | NP_001013267.1 | 10 | 1233 | ||
| XM_006243826.2 | XP_006243888.1 | 7 | 2360 | |||
| XM_006243827.2 | XP_006243889.1 | 9 | 1121 | |||
| XM_006243829.2 | XP_006243891.1 | 6 | 2249 | |||
| XM_006243830.1 | XP_006243892.1 | 8 | 1443 | |||
| XM_006243831.1 | XP_006243893.1 | 9 | 1089 | |||
| GenBank mRNA | BC091359.1 | - | 10 | 1233 | 7318733 | |

Dhx30
Genomic Map