siRNA Details
| Gene Symbol: Oard1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244466.2 | XP_006244528.1 | 5 | 916 | ||
| XM_006244467.2 | XP_006244529.1 | 5 | 913 | |||
| XM_006244470.2 | XP_006244532.1 | 5 | 508 | |||
| XM_006244471.2 | XP_006244533.1 | 5 | 741 | |||
| XM_006244472.2 | XP_006244534.1 | 5 | 512 | |||
| XM_008766872.1 | XP_008765094.1 | 5 | 924 | |||
| XM_008766873.1 | XP_008765095.1 | 5 | 922 | |||
| XM_008766874.1 | XP_008765096.1 | 6 | 788 | |||
| XM_008766875.1 | XP_008765097.1 | 5 | 646 | |||

Oard1
Genomic Map