siRNA Details
| Gene Symbol: Fam135a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134597.1 | NP_001128069.1 | 15 | 2021 | ||
| XM_006244664.1 | XP_006244726.1 | 14 | 2058 | |||
| XM_006244665.1 | XP_006244727.1 | 16 | 1729 | |||
| XM_006244666.1 | XP_006244728.1 | 13 | 1392 | |||
| GenBank mRNA | FQ222914.1 | - | 16 | 1729 | ||

Fam135a
Genomic Map