siRNA Details
Gene Symbol: Mgat4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012225.2 | NP_001012225.1 | 3, 4 | 535 | ||
GenBank mRNA | BC088215.1 | - | 2, 3 | 493 | 7383944 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mgat4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012225.2 | NP_001012225.1 | 3, 4 | 535 | ||
GenBank mRNA | BC088215.1 | - | 2, 3 | 493 | 7383944 |