siRNA Details
Gene Symbol: Dis3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109007.2 | NP_001102477.2 | 7 | 678 | ||
NM_001172159.1 | NP_001165630.1 | 6 | 636 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dis3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109007.2 | NP_001102477.2 | 7 | 678 | ||
NM_001172159.1 | NP_001165630.1 | 6 | 636 |