siRNA Details
| Gene Symbol: Gpr35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037359.1 | NP_001032436.1 | 1 | 67 | ||
| XM_006245537.2 | XP_006245599.1 | 2 | 210 | |||
| XM_006245539.2 | XP_006245601.1 | 2 | 425 | |||
| XM_006245540.2 | XP_006245602.1 | 2 | 422 | |||
| GenBank mRNA | AB240684.1 | - | 1 | 67 | ||

Gpr35
Genomic Map