siRNA Details
| Gene Symbol: Nudt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109010.1 | NP_001102480.1 | 6 | 1367 | ||
| XM_006245588.2 | XP_006245650.1 | 6 | 1372 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nudt12| Gene Symbol: Nudt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109010.1 | NP_001102480.1 | 6 | 1367 | ||
| XM_006245588.2 | XP_006245650.1 | 6 | 1372 | |||