siRNA Details
| Gene Symbol: Clul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033071.1 | NP_001028243.1 | 2 | 138 | ||
| GenBank mRNA | AY655707.1 | - | 2 | 138 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clul1| Gene Symbol: Clul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033071.1 | NP_001028243.1 | 2 | 138 | ||
| GenBank mRNA | AY655707.1 | - | 2 | 138 | ||