siRNA Details
| Gene Symbol: Pla2g12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006256432.2 | XP_006256494.1 | 1, 2 | 363 | ||
| XM_346131.7 | XP_346132.5 | 1, 2 | 362 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g12b| Gene Symbol: Pla2g12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006256432.2 | XP_006256494.1 | 1, 2 | 363 | ||
| XM_346131.7 | XP_346132.5 | 1, 2 | 362 | |||