siRNA Details
| Gene Symbol: EIF1AY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004681.3 | NP_004672.2 | 4, 5 | 415 | ||
| GenBank mRNA | AF000987.1 | - | 4, 5 | 386 | ||
| BC005248.1 | - | 4, 5 | 297 | 3933718 | ||
| BF694967.1 | - | 401 | ||||
| BG499107.1 | - | 4, 5 | 408 | |||
| BT007201.1 | - | 4, 5 | 254 | |||

EIF1AY
Genomic Map