siRNA Details
| Gene Symbol: Praf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109013.1 | NP_001102483.1 | 3 | 628 | ||
| GenBank mRNA | FQ212649.1 | - | 3 | 487 | ||
| FQ213026.1 | - | 3 | 487 | |||
| FQ213646.1 | - | 3 | 484 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Praf2| Gene Symbol: Praf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109013.1 | NP_001102483.1 | 3 | 628 | ||
| GenBank mRNA | FQ212649.1 | - | 3 | 487 | ||
| FQ213026.1 | - | 3 | 487 | |||
| FQ213646.1 | - | 3 | 484 | |||