siRNA Details
| Gene Symbol: Tceanc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109015.1 | NP_001102485.1 | 1 | 80 | ||
| XM_006256856.2 | XP_006256918.1 | 5 | 954 | |||
| XM_008773182.1 | XP_008771404.1 | 5 | 391 | |||
| XM_008773183.1 | XP_008771405.1 | 5 | 644 | |||

Tceanc
Genomic Map