siRNA Details
Gene Symbol: LOC367830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037211.1 | NP_001032288.1 | 2 | 425 | ||
GenBank mRNA | BC100149.1 | - | 2 | 425 | 7442108 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC367830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037211.1 | NP_001032288.1 | 2 | 425 | ||
GenBank mRNA | BC100149.1 | - | 2 | 425 | 7442108 |