siRNA Details
| Gene Symbol: Btk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007798.1 | NP_001007799.1 | 1581 | |||
| GenBank mRNA | AY787790.1 | - | 1581 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btk| Gene Symbol: Btk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007798.1 | NP_001007799.1 | 1581 | |||
| GenBank mRNA | AY787790.1 | - | 1581 | |||