siRNA Details
| Gene Symbol: Rbmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113786.2 | NP_001107258.2 | 5 | 581 | ||
| GenBank mRNA | BC158684.1 | - | 5 | 581 | 8373764 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbmx2| Gene Symbol: Rbmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113786.2 | NP_001107258.2 | 5 | 581 | ||
| GenBank mRNA | BC158684.1 | - | 5 | 581 | 8373764 | |