siRNA Details
Gene Symbol: LOC100912608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236523.2 | XP_006236585.1 | 6118 | |||
XM_006236524.2 | XP_006236586.1 | 6031 |
Gene Symbol: Hoxa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762948.1 | XP_008761170.1 | 1 | 6106 | ||
XM_008762949.1 | XP_008761171.1 | 2 | 655 |
Gene Symbol: LOC100911668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257709.2 | XP_006257771.1 | 1 | 6106 | ||
XM_008773714.1 | XP_008771936.1 | 2 | 655 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
