siRNA Details
| Gene Symbol: Ankib1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134781.1 | NP_001128253.1 | 12 | 1988 | ||
| XM_006236049.2 | XP_006236111.1 | 12 | 2240 | |||
| XM_006236050.2 | XP_006236112.1 | 13 | 3094 | |||
| XM_006236051.2 | XP_006236113.1 | 12 | 1690 | |||
| XM_008762709.1 | XP_008760931.1 | 12 | 3212 | |||
| XM_008762710.1 | XP_008760932.1 | 13 | 2606 | |||
| XM_008762711.1 | XP_008760933.1 | 12 | 3212 | |||

Ankib1
Genomic Map