siRNA Details
| Gene Symbol: Dgkd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245504.2 | XP_006245566.2 | 22 | 2704 | ||
| XM_006245505.2 | XP_006245567.2 | 22 | 2704 | |||
| GenBank mRNA | FQ218774.1 | - | 2 | 203 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dgkd| Gene Symbol: Dgkd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245504.2 | XP_006245566.2 | 22 | 2704 | ||
| XM_006245505.2 | XP_006245567.2 | 22 | 2704 | |||
| GenBank mRNA | FQ218774.1 | - | 2 | 203 | ||